siRNA Details
Gene Symbol: 2300009A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027090.1 | NP_081366.1 | 1, 2 | 373 | ||
GenBank mRNA | AK009046.1 | - | 1, 2 | 372 | ||
BC100363.1 | - | 1, 2 | 348 | |||
BY090300.1 | - | 1, 2 | 116 |
