siRNA Details
Gene Symbol: 2300009A05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027090.1 | NP_081366.1 | 2 | 452 | ||
GenBank mRNA | AK009046.1 | - | 2 | 451 | ||
BC100363.1 | - | 2 | 427 | |||
BY090300.1 | - | 2 | 195 |

More Information
Other Species Targeted

Gene Ontology Categories:
