siRNA Details
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271089.2 | NP_001258018.1 | 1 | 290 | ||
NM_019283.3 | NP_062156.2 | 2 | 415 | |||
GenBank mRNA | AB015433.1 | - | 2 | 381 | ||
BC061989.1 | - | 1 | 273 | 5599495 | ||
CB747485.1 | - | 2 | 415 | |||
FQ148223.1 | - | 1 | 290 | |||
U59324.1 | - | 1 | 204 |
