siRNA Details
Gene Symbol: Cd164l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027152.1 | NP_081428.1 | 5, 6 | 676 | ||
GenBank mRNA | AK009888.1 | - | 5, 6 | 602 | ||
AK139809.1 | - | 5, 6 | 678 | |||
BC111824.1 | - | 5, 6 | 599 | 40055637 |
