siRNA Details
Gene Symbol: Cacna1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019294.2 | NP_062167.2 | 1 | 251 | ||
GenBank mRNA | AY029412.1 | - | 1 | 251 | ||
L15453.1 | - | 1 | 291 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cacna1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019294.2 | NP_062167.2 | 1 | 251 | ||
GenBank mRNA | AY029412.1 | - | 1 | 251 | ||
L15453.1 | - | 1 | 291 |