siRNA Details
Gene Symbol: Lrch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081255.1 | NP_001074724.1 | 21 | 2963 | ||
GenBank mRNA | AK134026.1 | - | 11 | 1595 | ||
AK155550.1 | - | 21 | 2961 | |||
BC151058.1 | - | 21 | 2941 | 9087971 | ||
BC157924.1 | - | 20 | 2833 | 9088097 | ||
BC172144.1 | - | 19 | 2683 | 9054814 |
