siRNA Details
Gene Symbol: Crk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019302.1 | NP_062175.1 | 2 | 304 | ||
XM_006246913.1 | XP_006246975.1 | 2 | 482 | |||
GenBank mRNA | D44481.1 | - | 2 | 304 | ||
FQ210506.1 | - | 2 | 487 |

More Information
Other Species Targeted

Gene Ontology Categories:
