siRNA Details
Gene Symbol: Flt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019306.1 | NP_062179.1 | 1, 2 | 308 | ||
XM_008768998.1 | XP_008767220.1 | 1, 2 | 718 | |||
GenBank mRNA | AF157595.1 | - | 1, 2 | 72 | ||
D28498.1 | - | 1, 2 | 308 |
