siRNA Details
Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019309.2 | NP_062182.1 | 1 | 379 | ||
GenBank mRNA | Z11548.1 | - | 399 | |||
Z11715.1 | - | 1 | 177 |

More Information
Other Species Targeted

Gene Ontology Categories:
