siRNA Details
Gene Symbol: Kcnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019313.1 | NP_062186.1 | 1 | 209 | ||
GenBank mRNA | AF000973.1 | - | 1 | 209 |
Gene Symbol: LOC103690116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771520.1 | XP_008769742.1 | 4015 | |||
XM_008771521.1 | XP_008769743.1 | 2093 |
Gene Symbol: Omd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031817.1 | NP_114005.1 | 2 | 1198 | ||
XM_008771551.1 | XP_008769773.1 | 2 | 2093 | |||
GenBank mRNA | AF104362.1 | - | 2 | 1198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
