siRNA Details
Gene Symbol: Mafb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019316.1 | NP_062189.1 | 1 | 809 | ||
GenBank mRNA | U56241.1 | - | 1 | 809 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010658, BC038256, L36435, AK143209, AK131926, and others.
Gene Ontology Categories:
