siRNA Details
Gene Symbol: Eva1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199210.1 | NP_001186139.1 | 5 | 1117 | ||
GenBank mRNA | AF358257.1 | - | 5 | 844 | ||
AK016443.1 | - | 7 | 1113 | |||
BC132305.1 | - | 5 | 1117 | 40130582 | ||
BC144751.1 | - | 4 | 919 | |||
BC157969.1 | - | 3 | 667 | 9007291 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
