siRNA Details
Gene Symbol: Tysnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272090.1 | NP_001259019.1 | 4 | 1750 | ||
NM_001272091.1 | NP_001259020.1 | 3 | 1660 | |||
NM_001272092.1 | NP_001259021.1 | 3 | 1605 | |||
NM_027912.1 | NP_082188.1 | 4 | 1791 | |||
GenBank mRNA | AK005069.1 | - | 4 | 1792 | ||
AK154513.1 | - | 4 | 1774 | |||
BC119320.1 | - | 4 | 1769 | 8734069 | ||
BE847683.1 | - | 2 | 284 |
