siRNA Details
Gene Symbol: 1700007B14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164235.1 | NP_001157707.1 | 14 | 2060 | ||
NM_027944.1 | NP_082220.1 | 13 | 1942 | |||
GenBank mRNA | AK077102.1 | - | 15 | 1799 | ||
BC117767.1 | - | 14 | 1963 | 40090978 | ||
BC117768.1 | - | 13 | 1845 | 40090984 |
