siRNA Details
Gene Symbol: Fam219a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159583.1 | NP_001153055.1 | 6 | 1953 | ||
NM_027993.3 | NP_082269.2 | 6 | 1920 | |||
GenBank mRNA | AK009522.1 | - | 6 | 1904 | ||
AK134737.1 | - | 6 | 1959 |
