siRNA Details
Gene Symbol: P2rx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019256.1 | NP_062129.1 | 3 | 466 | ||
XM_008769223.1 | XP_008767445.1 | 3 | 640 | |||
GenBank mRNA | FJ436445.1 | - | 3 | 504 | ||
X95882.1 | - | 3 | 466 |
