siRNA Details
Gene Symbol: Lmntd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028050.2 | NP_082326.1 | 1 | 148 | ||
GenBank mRNA | BC119093.1 | - | 1 | 152 | 8733842 | |
BC119119.1 | - | 1 | 152 | 8733868 | ||
BC144834.1 | - | 1 | 152 | 9053382 |
