siRNA Details
Gene Symbol: Sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028182.1 | NP_082458.1 | 9 | 2412 | ||
GenBank mRNA | AK008803.1 | - | 9 | 2198 | ||
BC023784.1 | - | 9 | 1262 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sh2d4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028182.1 | NP_082458.1 | 9 | 2412 | ||
GenBank mRNA | AK008803.1 | - | 9 | 2198 | ||
BC023784.1 | - | 9 | 1262 |