siRNA Details
Gene Symbol: Kctd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289671.1 | NP_001276600.1 | 8 | 1488 | ||
NM_001289672.1 | NP_001276601.1 | 7 | 1413 | |||
NM_001289673.1 | NP_001276602.1 | 6 | 1325 | |||
GenBank mRNA | AK013500.1 | - | 1 | 340 |
