siRNA Details
Gene Symbol: 1700017B05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028820.2 | NP_083096.1 | 2 | 908 | ||
GenBank mRNA | AK029230.1 | - | 912 | |||
AK031730.1 | - | 2 | 911 | |||
AK044396.1 | - | 2 | 909 | |||
AK080676.1 | - | 2 | 920 | |||
AK155015.1 | - | 2 | 885 | |||
AK165963.1 | - | 2 | 909 | |||
BC118527.1 | - | 2 | 762 | 40098810 |
