siRNA Details
Gene Symbol: 1700017B05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028820.2 | NP_083096.1 | 3 | 3702 | ||
GenBank mRNA | AK029230.1 | - | 3706 | |||
AK044396.1 | - | 3 | 3710 | |||
AK155487.1 | - | 3 | 3703 | |||
AK165963.1 | - | 3 | 3703 | |||
AK169617.1 | - | 3 | 3705 | |||
BC046380.1 | - | 2 | 1344 | |||
BC118527.1 | - | 3 | 3556 | 40098810 |
