siRNA Details
Gene Symbol: Rhoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081105.1 | NP_001074574.1 | 3 | 4158 | ||
GenBank mRNA | AK137909.1 | - | 1 | 3588 | ||
BC040409.1 | - | 1 | 3533 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rhoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081105.1 | NP_001074574.1 | 3 | 4158 | ||
GenBank mRNA | AK137909.1 | - | 1 | 3588 | ||
BC040409.1 | - | 1 | 3533 |