siRNA Details
Gene Symbol: Meikin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB987828.1 | - | 13 | 1250 | ||
AK015073.1 | - | 13 | 1269 | |||
BC107368.1 | - | 12 | 1358 | 40056257 | ||
BC107369.1 | - | 13 | 1267 |
