siRNA Details
Gene Symbol: Morn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029112.1 | NP_083388.1 | 2 | 335 | ||
GenBank mRNA | AK077141.1 | - | 2 | 532 | ||
BC019987.1 | - | 2 | 335 | 4458628 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Morn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029112.1 | NP_083388.1 | 2 | 335 | ||
GenBank mRNA | AK077141.1 | - | 2 | 532 | ||
BC019987.1 | - | 2 | 335 | 4458628 |