siRNA Details
Gene Symbol: 4930519F16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029170.1 | NP_083446.1 | 4 | 780 | ||
GenBank mRNA | AK039711.1 | - | 4 | 780 | ||
BC107248.2 | - | 4 | 782 | 40052342 | ||
BC107249.1 | - | 4 | 778 | 40052344 | ||
BC114984.1 | - | 4 | 782 | 40052347 | ||
BC115753.1 | - | 4 | 780 | 40052345 |
