siRNA Details
Gene Symbol: 4930558K02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204904.1 | NP_001191833.1 | 6 | 519 | ||
NM_001204905.1 | NP_001191834.1 | 6 | 552 | |||
GenBank mRNA | AK016179.1 | - | 6 | 555 | ||
CF105154.1 | - | 6 | 519 |
