siRNA Details
Gene Symbol: 1700003F12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029305.2 | NP_083581.1 | 3 | 663 | ||
GenBank mRNA | AK005633.1 | - | 3 | 664 | ||
BC039797.1 | - | 3 | 525 | |||
BC147532.1 | - | 3 | 651 | 9056523 | ||
BC147533.1 | - | 3 | 651 | 9056524 | ||
DQ080431.1 | - | 3 | 664 |

More Information
Other Species Targeted

Gene Ontology Categories:
