siRNA Details
Gene Symbol: Them4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029431.1 | NP_083707.1 | 2 | 239 | ||
GenBank mRNA | AK014834.1 | - | 2 | 172 | ||
AK038420.1 | - | 2 | 137 | |||
AK138397.1 | - | 2 | 239 | |||
AK139993.1 | - | 2 | 242 | |||
BC022612.1 | - | 1 | 87 | |||
BC134396.1 | - | 2 | 235 | 40136254 |
