siRNA Details
Gene Symbol: Hormad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029458.1 | NP_083734.1 | 1 | 106 | ||
GenBank mRNA | BC120781.1 | - | 1 | 106 | 40129704 | |
BC120783.1 | - | 1 | 106 | 40129706 | ||
BC144842.1 | - | 1 | 106 | 9053390 |
