siRNA Details
Gene Symbol: Fam209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029608.1 | NP_083884.1 | 1 | 253 | ||
GenBank mRNA | AK006503.1 | - | 1 | 260 | ||
BC137932.1 | - | 1 | 253 | 8860951 | ||
BC137933.1 | - | 1 | 253 | 8860952 |
