siRNA Details
Gene Symbol: Cacna1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019294.2 | NP_062167.2 | 14, 15 | 1898 | ||
XM_006250069.2 | XP_006250131.1 | 16, 17 | 3215 | |||
GenBank mRNA | AY029412.1 | - | 14, 15 | 1898 | ||
L15453.1 | - | 14, 15 | 1938 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009782, BC150750.
Gene Ontology Categories:
