siRNA Details
Gene Symbol: 2700049A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163378.1 | NP_001156850.1 | 30 | 5604 | ||
NM_029818.1 | NP_084094.1 | 28 | 5340 | |||
GenBank mRNA | AK012399.1 | - | 13 | 2020 | ||
AK172976.1 | - | 28 | 4990 | |||
BC027577.1 | - | 2 | 219 | |||
BC043104.1 | - | 30 | 5604 | |||
BC046527.1 | - | 8 | 1271 |
