siRNA Details
Gene Symbol: 2700049A03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163378.1 | NP_001156850.1 | 2 | 1207 | ||
NM_029818.1 | NP_084094.1 | 2 | 1207 | |||
GenBank mRNA | AK040605.1 | - | 2 | 876 | ||
AK172976.1 | - | 2 | 858 | |||
BC043104.1 | - | 2 | 1207 | |||
BC150744.1 | - | 2 | 575 | 9087655 |
