siRNA Details
Gene Symbol: Inpp5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019311.1 | NP_062184.1 | 14, 15 | 1865 | ||
XM_008767293.1 | XP_008765515.1 | 12, 13 | 2445 | |||
GenBank mRNA | U55192.1 | - | 14, 15 | 1865 |
