siRNA Details
Gene Symbol: BUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278616.1 | NP_001265545.1 | 3 | 433 | ||
NM_001278617.1 | NP_001265546.1 | 4 | 493 | |||
NM_004336.4 | NP_004327.1 | 4 | 493 | |||
GenBank mRNA | AF043294.2 | - | 4 | 413 | ||
AF046078.1 | - | 4 | 430 | |||
AF047471.1 | - | 4 | 416 | |||
AF053305.1 | - | 4 | 444 | |||
AK023540.1 | - | 4 | 449 | |||
AK225317.1 | - | 4 | 449 | |||
AK291221.1 | - | 4 | 446 | |||
AK302418.1 | - | 3 | 389 | |||
BC017038.1 | - | 4 | 414 | |||
BC028201.1 | - | 4 | 414 | 5205385 | ||
BP313176.1 | - | 4 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
