siRNA Details
Gene Symbol: 9430038I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029886.2 | NP_084162.1 | 4 | 661 | ||
GenBank mRNA | AK078159.1 | - | 4 | 641 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9430038I01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029886.2 | NP_084162.1 | 4 | 661 | ||
GenBank mRNA | AK078159.1 | - | 4 | 641 |