siRNA Details
Gene Symbol: C330007P06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029951.1 | NP_084227.1 | 7 | 3105 | ||
GenBank mRNA | AK032304.1 | - | 7 | 3017 | ||
AK035825.1 | - | 7 | 3095 | |||
AK036910.1 | - | 7 | 3027 | |||
AK079769.1 | - | 1 | 1266 | |||
AK141615.1 | - | 7 | 3046 |
