siRNA Details
Gene Symbol: Msl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100451.2 | NP_001093921.1 | 815 | |||
GenBank mRNA | AK153696.1 | - | 1405 | |||
AK173208.1 | - | 1586 | ||||
BC070432.1 | - | 815 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Msl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100451.2 | NP_001093921.1 | 815 | |||
GenBank mRNA | AK153696.1 | - | 1405 | |||
AK173208.1 | - | 1586 | ||||
BC070432.1 | - | 815 |