siRNA Details
Gene Symbol: Crisp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030033.1 | NP_084309.1 | 7 | 938 | ||
GenBank mRNA | AY705449.1 | - | 7 | 938 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Crisp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030033.1 | NP_084309.1 | 7 | 938 | ||
GenBank mRNA | AY705449.1 | - | 7 | 938 |