siRNA Details
Gene Symbol: Crisp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030033.1 | NP_084309.1 | 5 | 488 | ||
GenBank mRNA | AK020340.1 | - | 6 | 549 | ||
AY705449.1 | - | 5 | 488 | |||
DQ200908.1 | - | 7 | 665 |
