siRNA Details
Gene Symbol: Cul9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081335.2 | NP_001074804.2 | 17 | 3897 | ||
GenBank mRNA | AK122351.1 | - | 14 | 2888 | ||
AK147675.1 | - | 16 | 4152 | |||
BC026469.1 | - | 2 | 151 |
