siRNA Details
Gene Symbol: 4930562C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030192.1 | NP_084468.1 | 10 | 2841 | ||
GenBank mRNA | BC137979.1 | - | 2 | 381 | ||
BC145075.1 | - | 2 | 370 | |||
BC145076.1 | - | 2 | 370 | |||
BC145794.1 | - | 2 | 381 |
