siRNA Details
Gene Symbol: Eif2s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019356.1 | NP_062229.1 | 2 | 165 | ||
GenBank mRNA | BC087019.1 | - | 2 | 222 | 7191037 | |
J02646.1 | - | 2 | 165 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026114, BC005463, AK010592, AK167763, BC016497, and others.
Gene Ontology Categories:
