siRNA Details
Gene Symbol: Pdpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019358.1 | NP_062231.1 | 2 | 356 | ||
XM_006239338.2 | XP_006239400.1 | 2 | 356 | |||
GenBank mRNA | BC072492.1 | - | 2 | 312 | 7097186 | |
U07797.1 | - | 2 | 356 | |||
U32115.1 | - | 2 | 284 | |||
U96449.1 | - | 2 | 136 |
