siRNA Details
Gene Symbol: Pdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271108.2 | NP_001258037.1 | 3 | 333 | ||
NM_019372.4 | NP_062245.2 | 2 | 328 | |||
XM_006237907.2 | XP_006237969.1 | 2 | 448 | |||
XM_006237908.2 | XP_006237970.1 | 2 | 321 | |||
XM_006237909.2 | XP_006237971.1 | 3 | 412 | |||
GenBank mRNA | AF062740.1 | - | 2 | 310 | ||
AW520836.1 | - | 3 | 333 | |||
BC129095.1 | - | 2 | 319 | 8369880 | ||
FN800686.1 | - | 2 | 328 |
