siRNA Details
Gene Symbol: PHKG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000294.2 | NP_000285.1 | 5 | 554 | ||
NM_001172432.1 | NP_001165903.1 | 5 | 554 | |||
GenBank mRNA | AK289492.1 | - | 5 | 418 | ||
AK293551.1 | - | 5 | 554 | |||
BC002541.2 | - | 5 | 414 | 3138927 | ||
M31606.1 | - | 5 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
