siRNA Details
Gene Symbol: Adam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020077.1 | NP_064462.1 | 3 | 197 | ||
GenBank mRNA | X99794.1 | - | 3 | 197 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020077.1 | NP_064462.1 | 3 | 197 | ||
GenBank mRNA | X99794.1 | - | 3 | 197 |