siRNA Details
Gene Symbol: NAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004536.2 | NP_004527.2 | 4 | 834 | ||
GenBank mRNA | AK309627.1 | - | 2 | 191 | ||
BC136273.1 | - | 2 | 171 | 9020260 | ||
BC143761.1 | - | 2 | 171 | 9052275 | ||
U19251.1 | - | 4 | 407 |
