siRNA Details
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019340.1 | NP_062213.1 | 13 | 2451 | ||
XM_006238260.2 | XP_006238322.1 | 20 | 3034 | |||
XM_008763800.1 | XP_008762022.1 | 9 | 1837 | |||
XM_008763801.1 | XP_008762023.1 | 4 | 1354 | |||
XM_008763802.1 | XP_008762024.1 | 4 | 2793 | |||
GenBank mRNA | AB055153.1 | - | 13 | 2451 |
