siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227853.2 | XP_006227915.1 | 28 | 4025 | ||
XM_008758714.1 | XP_008756936.1 | 27 | 4011 | |||
XM_008758715.1 | XP_008756937.1 | 29 | 4078 | |||
XM_008758716.1 | XP_008756938.1 | 29 | 4076 | |||
GenBank mRNA | AY597251.1 | - | 15 | 2421 | ||
X95466.1 | - | 14 | 2333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
